Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Chlorofuro[3,2-c]pyridine
4-Chlorofuro[3,2-c]pyridine
4-Chlorofuro[3,2-c]pyridine
Irritant
4-Chlorofuro[3,2-c]pyridine
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
4-Chlorofuro[3,2-c]pyridine
Supplier:
Apollo Scientific
About this item
CAS Number:
31270-80-1
:
Formula:
C₇H₄ClNO
MDL Number:
MFCD02179768
:
153.5658 g/mol
Melting Pt:
40…45 °C
:
GHS07
Storage Temperature:
Ambient