Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Furo[3,2-c]pyridin-4(5H)-one
Furo[3,2-c]pyridin-4(5H)-one
Furo[3,2-c]pyridin-4(5H)-one
:
Apollo Scientific
Furo[3,2-c]pyridin-4(5H)-one
:
Apollo Scientific
:
26956-43-4
:
:
C₇H₅NO₂
:
MFCD00183939
:
135.1201 g/mol
:
199…200 °C
:
GHS07
:
Ambient