Order Entry
Export
ContactUsLinkComponent
2,4-Quinolinediol
2,4-Quinolinediol
Catalog # APOSOR2578-100G
Supplier:  Apollo Scientific
CAS Number:  
2,4-Quinolinediol
Catalog # APOSOR2578-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :161.16 g/mol
Melting Pt:>300 °C
EINECS:201-711-0
 :GHS07
Storage Temperature:Ambient
CAS Number:86-95-3
 :70254-43-2
Formula:C₉H₇NO₂
MDL Number:MFCD00006744