0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
2,3-Dioxoindoline-7-carboxylic acid
2,3-Dioxoindoline-7-carboxylic acid
2,3-Dioxoindoline-7-carboxylic acid
APOSOR25737-1G
:
Apollo Scientific
:
2,3-Dioxoindoline-7-carboxylic acid
APOSOR25737-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
191.14 g/mol
:
268…275 °C
:
GHS07
:
Ambient
:
25128-35-2
:
C₉H₅NO₄
:
MFCD00612492