Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl 3,4-dimethylbenzoate
Methyl-3,4-dimethylbenzoate
Methyl-3,4-dimethylbenzoate
Irritant
Methyl-3,4-dimethylbenzoate
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
Methyl-3,4-dimethylbenzoate
Supplier:
Apollo Scientific
About this item
CAS Number:
38404-42-1
:
Formula:
C₁₀H₁₂O₂
MDL Number:
MFCD00219861
Melting Pt:
35…37 °C
:
164.2011 g/mol
:
GHS07
Storage Temperature:
Ambient