Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,6-Dichloro-4-methylpyridazine
3,6-Dichloro-4-methylpyridazine
3,6-Dichloro-4-methylpyridazine
3,6-Dichloro-4-methylpyridazine
APOSOR25616-1G
:
Apollo Scientific
:
3,6-Dichloro-4-methylpyridazine
APOSOR25616-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
163.01 g/mol
:
149…151 °C/21mmHg
:
86…88 °C
:
242-794-3
:
GHS07
:
Refrigerator
:
19064-64-3
:
C₅H₄Cl₂N₂
:
MFCD00006465