
2,6-Dichloro-4-nitrotoluene
Catalog # APOSOR25607-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:206.03 g/mol
Melting Pt:63…64 °C
:GHS07
Storage Temperature:Ambient
CAS Number:7149-69-1
Formula:C₇H₅Cl₂NO₂
MDL Number:MFCD00174267