
(1,4-Dimethylpiperazin-2-yl)methanol
Catalog # APOSOR25585-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:144.22 g/mol
Boiling Pt:114…115 °C/ 15mmHg
:GHS07
Storage Temperature:Ambient
CAS Number:14675-44-6
Formula:C₇H₁₆N₂O
MDL Number:MFCD00174342