Order Entry
Export
Orders LinkContactUsLinkComponent
 

4-Nitroimidazole

Irritant
Supplier:   Apollo Scientific
4-Nitroimidazole
Supplier:  Apollo Scientific
Certificates
4-Nitroimidazole

3 Options Available Below

Product Details & Documents
Synonyms:  4-Nitro-1H-imidazole, 5-Nitroimidazole, 5-Nitro-1H-imidazole
  • CAS Number:3034-38-6
  • Formula:C₃H₃N₃O₂
  • MDL Number:MFCD00005196
  • Molecular Weight:113.08 g/mol
  • Melting Pt:303 °C (dec.)
  • Flash Pt:>200 °C
  • Storage Temperature:Ambient
  • EINECS:221-224-7
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 21,80 €
    21,80 €
    1 * 25 g
       21,80 €

  • Bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 29,00 €
    29,00 €
    1 * 100 g
       29,00 €

  • Bottle
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 115,00 €
    115,00 €
    1 * 500 g
       115,00 €