1,3-Cyclopentanedione
Supplier: Apollo Scientific
Jump to:
About this item
Synonyms: Cyclopentane-1,3-dione
- CAS Number:3859-41-4
- Formula:C₅H₆O₂
- MDL Number:MFCD00001405
- Molecular Weight:98.1 g/mol
- Melting Pt:148…153 °C
- Storage Temperature:Refrigerator
2 Options Available Below
Product Details & Documents
Synonyms: Cyclopentane-1,3-dione
- CAS Number:3859-41-4
- Formula:C₅H₆O₂
- MDL Number:MFCD00001405
- Molecular Weight:98.1 g/mol
- Melting Pt:148…153 °C
- Storage Temperature:Refrigerator



