Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Hydroxy-1-tetralone
5-Hydroxy-1-tetralone 98%
5-Hydroxy-1-tetralone 98%
5-Hydroxy-1-tetralone 98%
Supplier:
Apollo Scientific
Certificates
Expand image
5-Hydroxy-1-tetralone 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
28315-93-7
:
Formula:
C₁₀H₁₀O₂
MDL Number:
MFCD00001693
:
162.19 g/mol
Melting Pt:
206…211 °C
Storage Temperature:
Ambient