Order Entry
Export
ContactUsLinkComponent
4,4'-Dimethylbiphenyl
Irritant
4,4'-Dimethylbiphenyl
Catalog # APOSOR24633-100G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4,4'-Dimethylbiphenyl
Catalog # APOSOR24633-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :182.27 g/mol
Boiling Pt:295 °C
Melting Pt:118…120 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:613-33-2
Formula:C₁₄H₁₄
MDL Number:MFCD00008545