Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-(1,3-Benzothiazol-2-yl)acetonitrile
2-(1,3-Benzothiazol-2-yl)acetonitrile 98%
2-(1,3-Benzothiazol-2-yl)acetonitrile 98%
Irritant
2-(1,3-Benzothiazol-2-yl)acetonitrile 98%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2-(1,3-Benzothiazol-2-yl)acetonitrile 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
56278-50-3
:
Formula:
C₉H₆N₂S
MDL Number:
MFCD00051633
:
174.23 g/mol
Melting Pt:
98…103 °C
:
GHS07
Storage Temperature:
Refrigerator