Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-Chloro[1,2,4]triazolo[4,3-b]pyridazine
6-Chloro[1,2,4]triazolo[4,3-b]pyridazine
6-Chloro[1,2,4]triazolo[4,3-b]pyridazine
APOSOR24235-250MG
:
Apollo Scientific
:
6-Chloro[1,2,4]triazolo[4,3-b]pyridazine
APOSOR24235-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
:
:
154.56 g/mol
:
202…210 °C
:
GHS07
:
Refrigerator
:
28593-24-0
:
C₅H₃ClN₄
:
MFCD00219047