Dicyclopropyl ketone 95%
Bottle, 25 mL
Catalog # APOSOR24134-25ML
Supplier: Apollo Scientific
Dicyclopropyl ketone 95%
Catalog # APOSOR24134-25ML
Supplier: Apollo Scientific
CAS Number: 1121-37-5
Jump to:
Product Details & Documents
Synonyms: Dicyclopropylketone, Dicyclopropylmethanone
- CAS Number:1121-37-5
- Formula:C₇H₁₀O
- MDL Number:MFCD00001276
- Molecular Weight:110.16 g/mol
- Boiling Pt:160…162 °C
- Density:0.968 g/cm³ (25 °C)
- Flash Pt:39 °C
- Storage Temperature:Ambient
- EINECS:214-331-5
- UN:1224
- ADR:3,III
Specifications
Specifications
- Pack type:Bottle
- Pk:25 mL



