Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Iodo-m-xylene
4-Iodo-m-xylene 98%
4-Iodo-m-xylene 98%
:
Apollo Scientific
4-Iodo-m-xylene 98%
:
Apollo Scientific
:
4214-28-2
:
:
C₈H₉I
:
MFCD00013706
:
105…108 °C/5mmHg
:
1.628 g/cm³ (25 °C)
:
>110 °C
:
232.06 g/mol
:
224-141-4
:
GHS07
:
Refrigerator