Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-(1,3-Oxazol-5-yl)aniline
4-(1,3-Oxazol-5-yl)aniline
4-(1,3-Oxazol-5-yl)aniline
Irritant
4-(1,3-Oxazol-5-yl)aniline
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
4-(1,3-Oxazol-5-yl)aniline
Supplier:
Apollo Scientific
About this item
CAS Number:
1008-95-3
:
Formula:
C₉H₈N₂O
MDL Number:
MFCD00052192
Melting Pt:
148…151 °C
:
160.1700 g/mol
:
GHS07
Storage Temperature:
Ambient