Order Entry
Export
ContactUsLinkComponent
 
2,6-Dimethoxybenzaldehyde 98%
Catalog # APOSOR24023-100G
Supplier:  Apollo Scientific
CAS Number:  
2,6-Dimethoxybenzaldehyde 98%
Catalog # APOSOR24023-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :166.18 g/mol
Boiling Pt:285 °C
Melting Pt:96…98 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:3392-97-0
Formula:C₉H₁₀O₃
MDL Number:MFCD00010862