Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-(1,3-Benzodioxol-5-yl)cyclohexane-1,3-dione
5-(1,3-Benzodioxol-5-yl)cyclohexane-1,3-dione
5-(1,3-Benzodioxol-5-yl)cyclohexane-1,3-dione
5-(1,3-Benzodioxol-5-yl)cyclohexane-1,3-dione
Supplier:
Apollo Scientific
Certificates
Expand image
5-(1,3-Benzodioxol-5-yl)cyclohexane-1,3-dione
Supplier:
Apollo Scientific
About this item
CAS Number:
55579-76-5
:
Formula:
C₁₃H₁₂O₄
MDL Number:
MFCD00111711
:
232.24 g/mol
Storage Temperature:
Ambient