
5,6-Dichloro-2-methylbenzimidazole
Catalog # APOSOR2383-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:201.0500 g/mol
Melting Pt:248 °C
:GHS07
Storage Temperature:Ambient
CAS Number:6478-79-1
Formula:C₈H₆Cl₂N₂
MDL Number:MFCD00276973