

1-Bromo-3,3-dimethylbutanone 97+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:5469-26-1
:
Formula:C₆H₁₁BrO
MDL Number:MFCD00000206
:179.06 g/mol
Boiling Pt:189…192 °C
Melting Pt:–10 °C
Density:1.333 g/cm³ (25 °C)
Flash Pt:75 °C
Storage Temperature:Refrigerator
UN:2810
ADR:6.1,II
:GHS05
:GHS06