
2-(2,6-Dichlorobenzylsulphanyl)ethylamine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:48133-71-7
:
Formula:C₉H₁₁Cl₂NS
MDL Number:MFCD00052684
Boiling Pt:152…154 °C/0.7mmHg
UN:3259
ADR:8,III
:236.16 g/mol
:GHS05
Storage Temperature:Ambient