
2,3-Dichlorobenzonitrile
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:6574-97-6
:
Formula:C₇H₃Cl₂N
MDL Number:MFCD00016372
:172.01 g/mol
Boiling Pt:114 °C/6mmHg
Melting Pt:60…64 °C
Storage Temperature:Ambient
UN:3439
ADR:6.1,III
:GHS07