Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,6-Dimethoxybenzyl alcohol
2,6-Dimethoxybenzyl alcohol
2,6-Dimethoxybenzyl alcohol
Irritant
2,6-Dimethoxybenzyl alcohol
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2,6-Dimethoxybenzyl alcohol
Supplier:
Apollo Scientific
About this item
CAS Number:
16700-55-3
:
Formula:
C₉H₁₂O₃
MDL Number:
MFCD00016862
:
168.19 g/mol
:
GHS07
Storage Temperature:
Ambient