

1,10-Phenanthroline hydrochloride monohydrate 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:18851-33-7
:3829-86-5
Formula:C₁₂H₉ClN₂·1H₂O
MDL Number:MFCD00150061
:234.68 g/mol
Melting Pt:224…225 °C (dec.)
UN:2811
ADR:6.1,III
:GHS06
:GHS09
Storage Temperature:Ambient