Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-Chloro-8-(chloromethyl)-4H-1,3-benzodioxine
6-Chloro-8-(chloromethyl)-4H-1,3-benzodioxine
6-Chloro-8-(chloromethyl)-4H-1,3-benzodioxine
APOSOR23581-1G
:
Apollo Scientific
:
6-Chloro-8-(chloromethyl)-4H-1,3-benzodioxine
APOSOR23581-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
219.07 g/mol
:
99…101 °C
:
GHS07
:
Ambient
:
175136-61-5
:
C₉H₈Cl₂O₂
:
MFCD00053071