4-Methoxybenzonitrile 99%
Supplier: Apollo Scientific
Jump to:
About this item
Synonyms: 4-Cyanoanisol, Anisonitrile, p-Cyanoanisol, p-Methoxybenzonitrile
- CAS Number:874-90-8
- Formula:C₈H₇NO
- MDL Number:MFCD00001818
- Molecular Weight:133.15 g/mol
- Boiling Pt:256…257 °C/765mmHg
- Melting Pt:57…60 °C
- Storage Temperature:Ambient
- EINECS:212-871-6
3 Options Available Below
Product Details & Documents
Synonyms: 4-Cyanoanisol, Anisonitrile, p-Cyanoanisol, p-Methoxybenzonitrile
- CAS Number:874-90-8
- Formula:C₈H₇NO
- MDL Number:MFCD00001818
- Molecular Weight:133.15 g/mol
- Boiling Pt:256…257 °C/765mmHg
- Melting Pt:57…60 °C
- Storage Temperature:Ambient
- EINECS:212-871-6



