
3,4-Dimethoxyphenol 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2033-89-8
:
Formula:C₈H₁₀O₃
MDL Number:MFCD00008390
:154.17 g/mol
Boiling Pt:167 °C/14mmHg
Melting Pt:79…82 °C
Storage Temperature:Ambient
EINECS:217-995-4
:GHS07