
4-(Chloromethyl)-2-phenyl-1,3-thiazole
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:4771-31-7
:
Formula:C₁₀H₈ClNS
MDL Number:MFCD00723503
:209.7 g/mol
Melting Pt:49…50 °C
Storage Temperature:Refrigerator
UN:3261
ADR:8,III
:GHS05