Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Phenyl-1,3-thiazole-4-carbaldehyde
2-Phenyl-1,3-thiazole-4-carbaldehyde
2-Phenyl-1,3-thiazole-4-carbaldehyde
Irritant
2-Phenyl-1,3-thiazole-4-carbaldehyde
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2-Phenyl-1,3-thiazole-4-carbaldehyde
Supplier:
Apollo Scientific
About this item
CAS Number:
20949-81-9
:
Formula:
C₁₀H₇NOS
MDL Number:
MFCD02681934
:
189.24 g/mol
Melting Pt:
51…52 °C
:
GHS07
Storage Temperature:
Refrigerator