Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Bromopyrimidine
2-Bromopyrimidine
2-Bromopyrimidine
:
Apollo Scientific
2-Bromopyrimidine
:
Apollo Scientific
:
4595-60-2
:
:
C₄H₃BrN₂
:
MFCD00014601
:
62…64 °C/1.5mmHg
:
53…57 °C
:
158.99 g/mol
:
Freezer
:
224-993-7
:
GHS07