Order Entry
Export
Orders LinkContactUsLinkComponent
2-Bromopyridine

2-Bromopyridine

Toxic
Supplier:   Apollo Scientific
2-Bromopyridine
Supplier:  Apollo Scientific
Certificates

3 Options Available Below

Product Details & Documents
  • CAS Number:109-04-6
  • Formula:C₅H₄BrN
  • MDL Number:MFCD00006219
  • Molecular Weight:158 g/mol
  • Boiling Pt:192…194 °C
  • Density:1.657 g/cm³ (25 °C)
  • Flash Pt:54 °C
  • Storage Temperature:Ambient
  • UN:2810
  • ADR:6.1,II
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    50 g
    Log in for availability
    Log in for availability
    1 * 50 g
    1 * 50 g - 21,80 €
    21,80 €
    1 * 50 g
       21,80 €

  • Bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 36,20 €
    36,20 €
    1 * 100 g
       36,20 €

  • Bottle
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 268,00 €
    268,00 €
    1 * 1 kg
       268,00 €