
2,3-Dihydrobenzo[1,4]dioxine-6-sulphonyl chloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:63758-12-3
:
Formula:C₈H₇ClO₄S
MDL Number:MFCD02677681
:234.66 g/mol
Melting Pt:65…71 °C
Storage Temperature:Ambient
UN:3261
ADR:8,II
:GHS05