Order Entry
Export
ContactUsLinkComponent
3,5-Dimethyl-4-isoxazolecarbaldehyde
Irritant
3,5-Dimethyl-4-isoxazolecarbaldehyde
Catalog # APOSOR22987-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
3,5-Dimethyl-4-isoxazolecarbaldehyde
Catalog # APOSOR22987-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :125.13 g/mol
Boiling Pt:54…56 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:54593-26-9
Formula:C6H7NO2
MDL Number:MFCD02681977