
3,5-Dimethylisoxazol-4-amine
Catalog # APOSOR22972-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
Melting Pt:41…46 °C
:112.13 g/mol
:GHS07
Storage Temperature:Freezer
CAS Number:31329-64-3
Formula:C₅H₈N₂O
MDL Number:MFCD02681972