Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Chloromethyl-1,4-benzodioxane
2-Chloromethyl-1,4-benzodioxane
2-Chloromethyl-1,4-benzodioxane
APOSOR22971-1G
:
Apollo Scientific
:
2-Chloromethyl-1,4-benzodioxane
APOSOR22971-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
184.62 g/mol
:
96 °C/0.4mmHg
:
GHS07
:
Ambient
:
2164-33-2
:
C₉H₉ClO₂
:
MFCD00014666