3,4-(Methylenedioxy)thiobenzamide
Catalog # APOSOR22962-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:181.21 g/mol
Storage Temperature:Ambient
CAS Number:15884-65-8
Formula:C₈H₇NO₂S
MDL Number:MFCD02677739