0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
2,2-Dimethylcyclopropanecarboxamide
2,2-Dimethylcyclopropanecarboxamide
2,2-Dimethylcyclopropanecarboxamide
Irritant
2,2-Dimethylcyclopropanecarboxamide
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2,2-Dimethylcyclopropanecarboxamide
Supplier:
Apollo Scientific
About this item
CAS Number:
1759-55-3
:
Formula:
C₆H₁₁NO
MDL Number:
MFCD00829406
:
113.16 g/mol
Melting Pt:
176…177 °C
:
GHS07
Storage Temperature:
Ambient