Order Entry
Export
ContactUsLinkComponent
 
Methyl picolinate 96%
Catalog # APOSOR22699-100G
Supplier:  Apollo Scientific
CAS Number:  
Methyl picolinate 96%
Catalog # APOSOR22699-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :137.14 g/mol
Storage Temperature:Ambient
CAS Number:2459-07-6
Formula:C₇H₇NO₂
MDL Number:MFCD00038038