2,6-Dibromoisonicotinic acid
Supplier: Apollo Scientific
Jump to:
About this item
Synonyms: 2,6-Dibromopyridine-4-carboxylic acid
- CAS Number:2016-99-1
- Formula:C₆H₃Br₂NO₂
- MDL Number:MFCD09743987
- Molecular Weight:280.9000 g/mol
- Storage Temperature:Refrigerator
3 Options Available Below
Product Details & Documents
Synonyms: 2,6-Dibromopyridine-4-carboxylic acid
- CAS Number:2016-99-1
- Formula:C₆H₃Br₂NO₂
- MDL Number:MFCD09743987
- Molecular Weight:280.9000 g/mol
- Storage Temperature:Refrigerator



