0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
2,6-Bis(hydroxymethyl)-p-cresol
2,6-Bis(hydroxymethyl)-p-cresol 95%
2,6-Bis(hydroxymethyl)-p-cresol 95%
APOSOR22124-500MG
:
Apollo Scientific
:
2,6-Bis(hydroxymethyl)-p-cresol 95%
APOSOR22124-500MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
500 mg
Seq:
2725
:
:
168.19 g/mol
:
126…130 °C
:
GHS07
:
Ambient
:
91-04-3
:
C₉H₁₂O₃
:
MFCD00004619