2,6-Dimethyl-4-hydrazinopyrimidine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:14331-56-7
:
Formula:C₆H₁₀N₄
MDL Number:MFCD00102150
Melting Pt:189…190 °C
UN:2811
ADR:6.1,III
:138.17 g/mol
:GHS07
Storage Temperature:Refrigerator