Order Entry
Export
ContactUsLinkComponent
4-Chloro-2,6-diaminopyrimidine
Irritant
4-Chloro-2,6-diaminopyrimidine
Catalog # APOSOR22082-25G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4-Chloro-2,6-diaminopyrimidine
Catalog # APOSOR22082-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :144.56 g/mol
Melting Pt:199…202 °C
EINECS:205-863-9
 :GHS07
Storage Temperature:Ambient
CAS Number:156-83-2
Formula:C₄H₅ClN₄
MDL Number:MFCD00006097