Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,2-Dimethylglutaric acid
2,2-Dimethylglutaric acid
2,2-Dimethylglutaric acid
Irritant
2,2-Dimethylglutaric acid
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2,2-Dimethylglutaric acid
Supplier:
Apollo Scientific
About this item
CAS Number:
681-57-2
:
Formula:
C₇H₁₂O₄
MDL Number:
MFCD00004197
:
160.17 g/mol
Boiling Pt:
200 °C
Melting Pt:
86 °C
:
GHS07
Storage Temperature:
Ambient