Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-(2,4-Dihydroxyphenyl)-2-methoxyethanone
1-(2,4-Dihydroxyphenyl)-2-methoxyethanone
1-(2,4-Dihydroxyphenyl)-2-methoxyethanone
Irritant
1-(2,4-Dihydroxyphenyl)-2-methoxyethanone
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
1-(2,4-Dihydroxyphenyl)-2-methoxyethanone
Supplier:
Apollo Scientific
About this item
CAS Number:
57280-75-8
:
Formula:
C₉H₁₀O₄
MDL Number:
MFCD00100481
:
182.18 g/mol
:
GHS07
Storage Temperature:
Ambient