Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4'-Amino-2'-hydroxyacetophenone
4'-Amino-2'-hydroxyacetophenone
4'-Amino-2'-hydroxyacetophenone
Irritant
4'-Amino-2'-hydroxyacetophenone
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
4'-Amino-2'-hydroxyacetophenone
Supplier:
Apollo Scientific
About this item
CAS Number:
2476-29-1
:
Formula:
C₈H₉NO₂
MDL Number:
MFCD00100636
:
151.1600 g/mol
Melting Pt:
129…131 °C
:
GHS07
Storage Temperature:
Ambient