Order Entry
Export
Orders LinkContactUsLinkComponent
3,5-Dinitrobenzonitrile 98+%

3,5-Dinitrobenzonitrile 98+%

Irritant
New Product
Supplier:   Apollo Scientific
3,5-Dinitrobenzonitrile 98+%
Supplier:  Apollo Scientific
New Product
Certificates
3,5-Dinitrobenzonitrile 98+%

2 Options Available Below

Product Details & Documents
  • CAS Number:4110-35-4
  • Formula:C₇H₃N₃O₄
  • MDL Number:MFCD00007228
  • Molecular Weight:193.12 g/mol
  • Melting Pt:126…130 °C
  • Storage Temperature:Ambient
  • EINECS:223-889-9
  • UN:3439
  • ADR:6.1,III
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 21,80 €
    21,80 €
    1 * 1 g
       21,80 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 944,00 €
    944,00 €
    1 * 100 g
       944,00 €

    Specifications:
    New Product
    New Product