Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Chloro-3,4-dimethoxybenzoic acid
2-Chloro-3,4-dimethoxybenzoic acid
2-Chloro-3,4-dimethoxybenzoic acid
:
Apollo Scientific
2-Chloro-3,4-dimethoxybenzoic acid
:
Apollo Scientific
:
52009-53-7
:
:
C₉H₉ClO₄
:
MFCD00084944
:
216.62 g/mol
:
200…204 °C
:
GHS07
:
Ambient