Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4,5-Dimethylfurfural
4,5-Dimethylfurfural
4,5-Dimethylfurfural
APOSOR21730-1G
:
Apollo Scientific
:
4,5-Dimethylfurfural
APOSOR21730-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
124.14 g/mol
:
98…100 °C/12mmHg
:
1.016 g/cm³ (25 °C)
:
66 °C
:
GHS07
:
Refrigerator
:
52480-43-0
:
C₇H₈O₂
:
MFCD00085030