Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Chloro-3-iodophenol
4-Chloro-3-iodophenol
4-Chloro-3-iodophenol
Irritant
4-Chloro-3-iodophenol
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
4-Chloro-3-iodophenol
Supplier:
Apollo Scientific
About this item
CAS Number:
202982-72-7
:
Formula:
C₆H₄ClIO
MDL Number:
MFCD00070769
Melting Pt:
75…77 °C
:
254.45 g/mol
:
GHS07
Storage Temperature:
Ambient